Merck
  • Research on L-nucleosides. Synthesis and biological evaluation of a series of L- and D-2',3'-dideoxy-3'-[tris(methylthio)methyl]-beta-pentofuranosyl nucleosides.
Research on L-nucleosides. Synthesis and biological evaluation of a series of L- and D-2',3'-dideoxy-3'-[tris(methylthio)methyl]-beta-pentofuranosyl nucleosides.
Bioorganic & medicinal chemistry (2003-01-09)
Claudia Mugnaini, Maurizio Botta, Massimo Coletta, Federico Corelli, Federico Focher, Stefano Marini, Michela Lucia Renzulli, Annalisa Verri
ABSTRACT
Novel nucleoside analogues of both D and L enantiomeric series were prepared by coupling reaction between a 2',3'-dideoxy-3'-modified furanose moiety and four different nucleobases. Though in all cases anomeric mixtures of nucleosides were obtained, the presence of the sterically bulky 3'-tris(methylthio)methyl group allowed a good stereoselectivity level. All the compounds of both enantiomeric series showed high IC(50) values as HSV-1 TK inhibitors and scarce ability to be phosphorylated by HSV-1 TK. In order to overcome possible problems related to the first phosphorylation step and to facilitate the penetration of the molecule through the cellular membrane, a monophosphate prodrug containing a long lipophilic chain was synthesized. No appreciable antiviral activity was exhibited by this molecule.
Sigma-Aldrich
Tris(methylthio)methane, 98%