Merck
  • The degradation (by distinct pathways) of human D-amino acid oxidase and its interacting partner pLG72--two key proteins in D-serine catabolism in the brain.
The degradation (by distinct pathways) of human D-amino acid oxidase and its interacting partner pLG72--two key proteins in D-serine catabolism in the brain.
The FEBS journal (2013-11-19)
Pamela Cappelletti, Paola Campomenosi, Loredano Pollegioni, Silvia Sacchi
ABSTRACT
Human D-amino acid oxidase (EC 1.4.3.3; hDAAO) is a peroxisomal flavoenzyme significantly enriched in the mammalian brain. hDAAO has been proposed to play (with serine racemase; EC 5.1.1.18) an essential role in the catabolism of D-serine, an 'atypical' key signalling molecule that acts as allosteric activator of the N-methyl-D-aspartate-type glutamate receptor (NMDAr). hDAAO and its interacting partner pLG72 have been related to schizophrenia, a highly disabling psychiatric disorder in which a dysfunction of NMDA-mediated neurotransmission is widely assumed to occur. We previously demonstrated that the D-serine cellular concentration depends on hDAAO and pLG72 expression levels and that newly-synthesized hDAAO interacts with its modulator in the cytosol, being progressively destabilized and inactivated. To obtain insight into the mechanisms regulating cellular D-serine levels, we investigated the degradation pathways of hDAAO and pLG72 in U87 glioblastoma cells stably expressing enhanced yellow fluorescent protein-hDAAO (peroxisomal), hDAAO-enhanced yellow fluorescent protein (cytosolic) or pLG72-enhanced cyan fluorescent protein (mitochondrial) proteins. hDAAO is a long-lived protein: the peroxisomal fraction of this flavoprotein is degraded via the lysosomal/endosomal pathway (and blocking this pathway increases the cellular hDAAO activity and decreases D-serine levels), whereas the cytosolic portion is ubiquitinated and targeted to the proteasome. By contrast, pLG72 shows a rapid turnover (t(1/2) ≈ 25-40 min) and is degraded via the proteasome system, albeit not ubiquitinated. Overexpression of pLG72 increases the turnover of hDAAO, in turn playing a protective role against excessive D-serine depletion.
Sigma-Aldrich
D-Amino Acid Oxidase from porcine kidney, powder, ≥1.5 units/mg solid
Pricing and availability is not currently available.