Merck
  • Hyaluronan derivatives: Alkyl chain length boosts viscoelastic behavior to depolymerization.
Hyaluronan derivatives: Alkyl chain length boosts viscoelastic behavior to depolymerization.
Carbohydrate polymers (2013-08-06)
Mauro Pavan, Devis Galesso, Giampaolo Menon, Davide Renier, Cristian Guarise
ABSTRACT
Five amide derivatives of Hyaluronic Acid (HA) were synthesized with C8, C12, C15, C16 and C18 linear alkyl-amines. These polymers (Hyadd) were tested against thermal, oxidative and hyaluronidase degradation by means of rheological experiments and SEC analysis and compared to non-modified HA. First of all, no free hexadecylamine was detected in the treated samples, meaning that under these stressing conditions only cleavage of glycosidic bonds occurs. Then, viscoelastic properties were assessed during thermal degradation and their variation as a function of time was expressed by means of a decay constant k(G'): while no significant difference in the decrease rate was observed between Hyadd-C8 and Hyadd-C12, a marked stabilization of viscoelastic properties during thermal treatment was detected for Hyadd-C15, Hyadd-C16 and Hyadd-C18. On the other hand, no difference was observed between the MW decrease rate (kMW decay constant) of HA and Hyadd-C12 to-C18; the depolymerization takes place on the backbone of the polymers independently whether they are derivatized or not, but longer alkyl chains lead to higher viscoelasticity in the depolymerized products. Finally, both oxidative and enzymatic degradation were carried out analyzing the changes in elastic modulus and in dynamic viscosity: once again, the amide side chain came out with similar behavior to chemical cross-linked HA (HBC) and with improved performances respect to linear HA in terms of preservation of viscoelasticity after chain depolymerization.
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid