Merck
  • Dysfunctional CD39(POS) regulatory T cells and aberrant control of T-helper type 17 cells in autoimmune hepatitis.
Dysfunctional CD39(POS) regulatory T cells and aberrant control of T-helper type 17 cells in autoimmune hepatitis.
Hepatology (Baltimore, Md.) (2013-06-22)
Charlotte R Grant, Rodrigo Liberal, Beth S Holder, John Cardone, Yun Ma, Simon C Robson, Giorgina Mieli-Vergani, Diego Vergani, Maria Serena Longhi
ABSTRACT
Autoimmune hepatitis (AIH) is an important cause of severe liver disease and is associated with both quantitative and qualitative regulatory T-cell (Treg) impairments. Tregs express CD39, an ectonucleotidase responsible for extracellular nucleotide hydrolysis, culminating in the production of immunosuppressive adenosine. Here, we describe multiple CD39(pos) Treg defects that potentially contribute to the impaired immunoregulation that is characteristic of AIH. We have examined the frequency and phenotype of CD39(pos) Tregs by flow cytometry and measured their ectonucleotidase activity. The capacity of CD4(pos) CD25(high) , CD4(pos) CD25(high) CD39(pos) , and CD4(pos) CD25(high) CD39(neg) subsets to suppress both proliferation of effector T cells and interleukin (IL)-17 production was evaluated. In AIH, CD39(pos) Tregs are decreased in frequency, exhibit limited adenosine triphosphate/adenosine diphosphate hydrolysis activity, and fail to suppress IL-17 production by effector CD4 T cells. Moreover, these CD39(pos) Tregs display a more proinflammatory profile in AIH, which is characterized by elevated CD127 positivity, and a greater propensity to produce interferon-gamma or IL-17 upon challenge with proinflammatory stimuli. In AIH, CD39(pos) Tregs are decreased in number, fail to adequately hydrolyze proinflammatory nucleotides and do not efficiently suppress IL-17 production by effector CD4 T cells. CD39(pos) Tregs show plasticity and are unstable upon proinflammatory challenge, suggesting that defective immunoregulation in AIH might result not only from reduced Treg number and function, but also from increased conversion of Tregs into effector cells.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, High Activity, ATPase ≥600 units/mg protein, lyophilized powder