Merck
  • HLA-F and MHC-I open conformers cooperate in a MHC-I antigen cross-presentation pathway.
HLA-F and MHC-I open conformers cooperate in a MHC-I antigen cross-presentation pathway.
Journal of immunology (Baltimore, Md. : 1950) (2013-07-16)
Jodie P Goodridge, Ni Lee, Aura Burian, Chul-Woo Pyo, Scott S Tykodi, Edus H Warren, Cassian Yee, Stanley R Riddell, Daniel E Geraghty
ABSTRACT
Peptides that are presented by MHC class I (MHC-I) are processed from two potential sources, as follows: newly synthesized endogenous proteins for direct presentation on the surface of most nucleated cells and exogenous proteins for cross-presentation typically by professional APCs. In this study, we present data that implicate the nonclassical HLA-F and open conformers of MHC-I expressed on activated cells in a pathway for the presentation of exogenous proteins by MHC-I. This pathway is distinguished from the conventional endogenous pathway by its independence from TAP and tapasin and its sensitivity to inhibitors of lysosomal enzymes, and further distinguished by its dependence on MHC-I allotype-specific epitope recognition for Ag uptake. Thus, our data from in vitro experiments collectively support a previously unrecognized model of Ag cross-presentation mediated by HLA-F and MHC-I open conformers on activated lymphocytes and monocytes, which may significantly contribute to the regulation of immune system functions and the immune defense.
Sigma-Aldrich
Anti-HLA-F antibody produced in rabbit, affinity isolated antibody