Merck
  • Synthesis and antimicrobial activity of some cyclic and acyclic nucleosides of thieno[2,3-d]pyrimidines.
Synthesis and antimicrobial activity of some cyclic and acyclic nucleosides of thieno[2,3-d]pyrimidines.
Nucleosides, nucleotides & nucleic acids (2007-05-05)
Nasser A Hassan, Mohamed I Hegab, Aymn E Rashad, Afaf A Fahmy, Farouk M E Abdel-Megeid
ABSTRACT
The reaction of compounds 1, 2, 3, 4, or 13 with 2-chloroethyl methyl ether or 2,3,4,6-tetra-O-acetyl-alpha-D-glucopyranosyl bromide, afforded some acyclic and cyclic nucleosides of thieno[2,3-d]pyrimidine derivatives. Furthermore, cyclic C-nucleosides 24 and 25 were prepared from the reaction of 20, 21 or from 26, 27 with D-glucose. The antimicrobial evaluation of some prepared products showed promising antimicrobial activity.
Sigma-Aldrich
2-Chloroethyl methyl ether, 98%
Pricing and availability is not currently available.