Merck
Fibrous histiocytoma of limbus.
Acta ophthalmologica (1989-10-01)
S A Schellini, C E Bacchi
ABSTRACT
The authors report a case of fibrohistiocytoma of the limbus and discuss the clinical, histopathological and immunohistochemical findings concerning this type of lesion, with a comparison of their findings with those reported in the literature.
Sigma-Aldrich
Fructosyl-Amino Acid Oxidase from Corynebacterium sp., recombinant, expressed in E. coli, lyophilized powder, ≥0.45 units/mg protein