Merck
  • Nucleophilic dearomatization of chloromethyl naphthalene derivatives via η3-benzylpalladium intermediates: a new strategy for catalytic dearomatization.
Nucleophilic dearomatization of chloromethyl naphthalene derivatives via η3-benzylpalladium intermediates: a new strategy for catalytic dearomatization.
Organic letters (2011-09-14)
Bo Peng, Sheng Zhang, Xiaoqiang Yu, Xiujuan Feng, Ming Bao
ABSTRACT
Palladium-catalyzed nucleophilic dearomatization of chloromethyl naphthalene derivatives to produce ortho- or para-substituted carbocycles in satisfactory to excellent yields has been developed. The unprecedented dearomatization reactions proceeded smoothly under mild conditions via η(3)-benzylpalladium intermediates.
Sigma-Aldrich
1-(Chloromethyl)naphthalene, technical, ≥97.0% (GC)
Pricing and availability is not currently available.