Merck
  • Long-term observation of fluorescence of free single molecules to explore protein-folding energy landscapes.
Long-term observation of fluorescence of free single molecules to explore protein-folding energy landscapes.
Journal of the American Chemical Society (2012-06-14)
Kiyoto Kamagata, Toshifumi Kawaguchi, Yoshitomo Iwahashi, Akinori Baba, Kazuya Fujimoto, Tamiki Komatsuzaki, Yoshihiro Sambongi, Yuji Goto, Satoshi Takahashi
ABSTRACT
A method was developed to detect fluorescence intensity signals from single molecules diffusing freely in a capillary cell. A unique optical system based on a spherical mirror was designed to enable quantitative detection of the fluorescence intensity. Furthermore, "flow-and-stop" control of the sample can extend the observation time of single molecules to several seconds, which is more than 1000 times longer than the observation time available using a typical confocal method. We used this method to scrutinize the fluorescence time series of the labeled cytochrome c in the unfolded state. Time series analyses of the trajectories based on local equilibrium state analysis revealed dynamically differing substates on a millisecond time scale. This system presents a new avenue for experimental characterization of the protein-folding energy landscape.
Sigma-Aldrich
Atto 532 maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)
Sigma-Aldrich
Atto 532, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 532 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)