Merck
  • [Three-year follow-up of 12 patients with prostate cancer treated with monthly degarelix in a phase II clinical trial].
[Three-year follow-up of 12 patients with prostate cancer treated with monthly degarelix in a phase II clinical trial].
Gan to kagaku ryoho. Cancer & chemotherapy (2014-01-16)
Senji Hoshi, Natsuho Hayashi, Mayu Yagi, Teppei Ookubo, Akinori Muto, Osamu Sugano, Kenji Numahata, Vladimir Bilim, Kiyotugu Hoshi, Isoji Sasagawa
ABSTRACT
The efficacy and safety of degarelix, a luteinizing hormone-releasing hormone(LH-RH)antagonist, in patients with prostate cancer(PCa)were evaluated in a phase II, open-label, multicenter clinical trial. In this trial, a total of 13 patients were accrued at the Yamagata Prefectural Central Hospital from 2007 to 2008. The median age was 80 years(range, 65-85 years), and clinical stages were T1c, T2, T3, and T4 in 1, 4, 6, and 2 patients, respectively. Nodal(N)status was N0 in 9 patients and N1 in 4 patients. Distant metastases were absent(M0)in 12 patients and present(M1b)in 1 patient. The median prostate- specific antigen(PSA)level was 29.1 ng/mL(range, 6.3-427 ng/mL). All but one patient, who died of an unrelated cause, received a monthly dose(80 or 160mg)of degarelix for 12 months and were followed-up for 3 years. The PSA level declined in all patients. One patient died of an unrelated cause during the phase II trial. After completion of the phase II trial, 5 patients were treated with combined and rogen blockade(CAB)(leuprolide plus anti-androgen therapy), 2 patients were treated with single-agent leuprolide, 2 patients received single-agent bicalutamide, and 1 patient was followed-up without additional treatment. Radical prostatectomy was performed in 2 patients. Among the 5 patients treated with CAB, 2 died of metastatic cancer. CAB was effective in suppressing PSA levels in 3 patients. In 1 patient with T3aN1M1b PCa, colon cancer with lung metastases was detected during the follow-up period. Treatment with chemotherapy for colon cancer was effective in suppressing PSA levels for 12 months. In 1 patient with cT3aN1M0 PCa, the PSA level declined to <0.02 ng/mL, and a reduction in size of the prostate gland and metastatic lymph nodes was observed. This effect persisted for 3.5 years after the completion of the 12-month degarelix regimen, and no additional treatment was required.