Merck
  • Bone mineral density trends in Indian patients with hyperthyroidism--effect of antithyroid therapy.
Bone mineral density trends in Indian patients with hyperthyroidism--effect of antithyroid therapy.
The Journal of the Association of Physicians of India (2012-02-18)
Dinesh Kumar Dhanwal, Nandita Gupta
ABSTRACT
Hyperthyroidism is associated with bone loss, which is reversible after treatment. The extent of reversibility of loss of bone mass density (BMD) in hyperthyroid patients after treatment especially at forearm is not clear. Therefore, the present study was conducted to assess degree of reversibility in bone mineral density following one-year medical treatment in Indian patients with hyperthyroidism. A total of 30 consecutive patients with hyperthyroidism were included in this one year study at All India Institute of Medical Sciences, New Delhi, India. All the patients were assessed for parameters of bone mineral homeostasis such as calcium, phosphorous, alkaline phosphatase, 25-hydroxy vitamin D [25 (OH) D], parathyroid hormone (PTH) at the time of diagnosis and after one year medical treatment. Bone mineral density was measured using Hologic DXA scan at hip, spine and forearm. All the patients received medical therapy with carbimazole. The parameters of bone homeostasis and bone mineral density at base line and after one year medical treatment was compared. All patients attained euthyroid status after eight weeks of carbimazole therapy. Parameters of bone homeostasis such as calcium, phosphorous, 25 (OH) D and PTH did not show any significant change from base line. Bone mineral density expressed as bone mineral content in gm/cm2 at left hip neck, trochanteric and intertrochanteric region was significantly higher after carbimazole therapy (745.2 +/- 127.6 gm/cm2 vs. 688.2 +/- 123.5 gm/cm2; p = 0.02, 573.4 +/- 109.9 gm/cm2 vs. 641.0 +/- 138.0 gm/cm2, p = 0.005 and 1008.6 +/- 185.5 gm/cm2 vs. 938.0 +/- 145.3 gm/cm2 p = 0.0131 respectively). Bone mineral density at lumbar spine expressed as either T and Z score was significantly higher after treatment (10 months of euthyroid state) (-0.6 +/- 1.3 vs. -1.7 +/- 1.2, p = 0.013 and -0.4 +/- 1.2 vs. -1.4 +/- 1.2, p = 0.012 respectively). However Bone mineral measures as T and Z score at left forearm decreased significantly after one year of medical therapy. In Indian patients with hyperthyroidism, the pattern of recovery of bone loss after one year of antithyroid therapy suggests early recovery at hip and lumbar spine and deterioration at forearm.
Sigma-Aldrich
Carbimazole, ≥98% (HPLC)
Carbimazole, European Pharmacopoeia (EP) Reference Standard