Merck
  • Ampicillin concentrations in human radicular granuloma following a single oral dose of bacampicillin.
Ampicillin concentrations in human radicular granuloma following a single oral dose of bacampicillin.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (1992-01-01)
Y Akimoto, K Kaneko, A Fujii, H Yamamoto
ABSTRACT
Ampicillin concentrations in human serum and radicular granulomas of 42 patients were determined after a single oral dose of bacampicillin (equivalent to 500 mg of ampicillin). Although wide variations were found among both serum and radicular granuloma ampicillin concentrations, measurable concentrations were found in all cases. The mean peak ampicillin concentrations in serum and radicular granulomas occurred at identical times, 1.5 hours, and were 11.19 micrograms/mL (range, 1.30 to 21.00 micrograms/mL) and 5.12 micrograms/g (range, 0.50 to 10.50 micrograms/g), respectively. The mean radicular granuloma/serum ampicillin concentration ratio at the peak time was 0.42. Ampicillin concentrations in radicular granulomas exceeded most of the minimum inhibitory concentrations for bacteria commonly isolated from odontogenic infections.
Product Description
Bacampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.