Merck
  • PGK1 as predictor of CXCR4 expression, bone marrow metastases and survival in neuroblastoma.
PGK1 as predictor of CXCR4 expression, bone marrow metastases and survival in neuroblastoma.
PloS one (2014-01-01)
Helen M Ameis, Astrid Drenckhan, Katharina von Loga, Gabriele Escherich, Katharina Wenke, Jakob R Izbicki, Konrad Reinshagen, Stephanie J Gros
ABSTRACT
A close relationship between phosphoglycerate kinase 1 (PGK1) and the CXCR4/SDF1 axis (chemokine receptor 4/stromal cell derived factor 1) has been shown for several cancers. However, the role of PGK1 has not been investigated for neuroblastoma, and PGK1 might be a therapeutic target for this tumor entity. The aim of the current study was to evaluate the role of PGK1 expression in neuroblastoma patients, to determine the impact of PGK1 expression levels on survival, and to correlate PGK1 expression with CXCR4 expression and bone marrow dissemination. Samples from 22 patients with neuroblastoma that were surgically treated at the University Medical Center Hamburg-Eppendorf were evaluated for expression of PGK1 and CXCR4 using immunohistochemistry. Results were correlated with clinical parameters, metastases and outcome of patients. Immunocytochemistry, proliferation and expression analysis of CXCR4 and PGK1 were performed in neuroblastoma cell lines. PGK1 is expressed in neuroblastoma cells. PGK1 expression is significantly positively correlated with CXCR4 expression and tumor dissemination to the bone marrow. Moreover the expression of PGK1 is significantly associated with a negative impact on survival in patients with neuroblastoma. PGK1 is downregulated by inhibition of CXCR4 in neuroblastoma cells. PGK1 appears to play an important role for neuroblastoma, predicting survival and tumor dissemination. Further in vivo studies outstanding, it is a candidate target for novel therapeutic strategies.
Sigma-Aldrich
Anti-PGK1 (AB1) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.