Merck
C-C bond formation with carbon dioxide promoted by a silver catalyst.
Angewandte Chemie (International ed. in English) (2012-06-08)
Satoshi Kikuchi, Kohei Sekine, Tomonobu Ishida, Tohru Yamada
ABSTRACT
A catalytic amount of silver benzoate with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene (MTBD) was an effective catalytic system for the reaction of carbon dioxide with various ketones containing an alkyne group at an appropriate position. These reactions afforded the corresponding γ-lactone derivatives in good to high yields under mild conditions.
Sigma-Aldrich
Silver benzoate, 99%
Pricing and availability is not currently available.