Merck
The effect of camphor on sex hormones levels in rats.
Cell journal (2014-02-26)
Sima Shahabi, Seyed Gholam Ali Jorsaraei, Ali Akbar Moghadamnia, Effat Barghi, Ebrahim Zabihi, Masoumeh Golsorkhtabar Amiri, Ghorban Maliji, Alieh Sohan Faraji, Maryam Abdi Boora, Neda Ghazinejad, Hajar Shamsai
ABSTRACT
In some traditional therapies, it has been claimed that camphor (a crystalline ketone obtained from cinnamomum camphora) would be a suppressor of sexual behaviors and sex hormones. This study evaluated the effects of camphor on sex hormones, like luteinizing hormone (LH), follicle-stimulating hormone (FSH) and testosterone. In this experimental study, 56 male rats were divided into 5 groups, including control (n=12), sham (n=11) and three treatment groups (n=11) in three different doses. The sham groups received daily intra peritoneal (IP) injections of the vehicle (ethanol 10%) for 30 days. Three treatment groups received different daily IP injections of the camphor (1, 2 and 5 mg/Kg) for 30 days and the control groups didn't received anything. Serums were used for assaying LH, FSH and testosterone. The level of LH significantly increased in all doses of camphor among the treatment groups as compared to the control (p<0.05), but camphor in doses 2 and 5 mg/Kg significantly reduced the FSH level as compared to control group (p<0.05). No significant changes were seen in testosterone levels. Camphor increased level of LH, decreased level of FSH, whereas it failed to change level of testosterone. The claim of inhibitory effect of camphor on sexual activity could not be confirmed by this study. More investigations in this field are suggested.
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Pricing and availability is not currently available.