Merck
  • Clinical characteristics and long-term outcome of young hairy cell leukemia patients treated with cladribine: a single-institution series.
Clinical characteristics and long-term outcome of young hairy cell leukemia patients treated with cladribine: a single-institution series.
Blood (2013-11-07)
Joshua D Rosenberg, Carol Burian, Jill Waalen, Alan Saven
ABSTRACT
Hairy cell leukemia (HCL) is a rare, indolent B-cell disorder in which single courses of cladribine induce high rates of complete responses. We report on 88 young HCL patients (≤40 years of age at diagnosis) treated with cladribine from the Scripps Clinic HCL Database, of whom 83 were evaluable for response. Seventy-three patients (88%) achieved an initial complete response and 10 (12%) a partial response, with a median response duration of 57 months. Forty-eight patients (58%) relapsed, with a median time to first relapse for all responders of 54 months. Eight patients developed 11 second primary malignancies with an excess frequency of 1.60 (95% confidence interval, 0.80-2.89). Thirteen (15%) patients died with a mortality ratio compared with age-matched normals of 1.85 (95% confidence interval, 1.07-3.18). Median overall survival for all patients following the first cladribine course was 231 months, and 251 months from diagnosis. Single courses of cladribine induce high rates of complete and durable responses in the majority of young HCL patients and are therefore recommended for HCL patients regardless of age.
Sigma-Aldrich
2-Chloro-2′-deoxyadenosine, antileukemic