Merck
  • Plant sterols the better cholesterol in Alzheimer's disease? A mechanistical study.
Plant sterols the better cholesterol in Alzheimer's disease? A mechanistical study.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-10-11)
Verena K Burg, Heike S Grimm, Tatjana L Rothhaar, Sven Grösgen, Benjamin Hundsdörfer, Viola J Haupenthal, Valerie C Zimmer, Janine Mett, Oliver Weingärtner, Ulrich Laufs, Laus M Broersen, Heikki Tanila, Tim Vanmierlo, Dieter Lütjohann, Tobias Hartmann, Marcus O W Grimm
ABSTRACT
Amyloid-β (Aβ), major constituent of senile plaques in Alzheimer's disease (AD), is generated by proteolytic processing of the amyloid precursor protein (APP) by β- and γ-secretase. Several lipids, especially cholesterol, are associated with AD. Phytosterols are naturally occurring cholesterol plant equivalents, recently been shown to cross the blood-brain-barrier accumulating in brain. Here, we investigated the effect of the most nutritional prevalent phytosterols and cholesterol on APP processing. In general, phytosterols are less amyloidogenic than cholesterol. However, only one phytosterol, stigmasterol, reduced Aβ generation by (1) directly decreasing β-secretase activity, (2) reducing expression of all γ-secretase components, (3) reducing cholesterol and presenilin distribution in lipid rafts implicated in amyloidogenic APP cleavage, and by (4) decreasing BACE1 internalization to endosomal compartments, involved in APP β-secretase cleavage. Mice fed with stigmasterol-enriched diets confirmed protective effects in vivo, suggesting that dietary intake of phytosterol blends mainly containing stigmasterol might be beneficial in preventing AD.
Sigma-Aldrich
Stigmasterol, ~95%
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-BACE 1, C-Terminus (485-501) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.