Merck
  • Immune microenvironment profiles of tumor immune equilibrium and immune escape states of mouse sarcoma.
Immune microenvironment profiles of tumor immune equilibrium and immune escape states of mouse sarcoma.
Cancer letters (2013-08-08)
Xi Wu, Meiyu Peng, Bingqing Huang, Huan Zhang, Huafeng Wang, Biao Huang, Zhenyi Xue, Lijuan Zhang, Yurong Da, De Yang, Zhi Yao, Rongxin Zhang
ABSTRACT
Cancer immunoediting consists of three distinct phases: elimination, equilibrium and escape. Here, for the first time, we investigated the immune microenvironment profiles of tumor immune equilibrium and immune escape states in 3'-methylcholanthrene-induced murine sarcoma model. Our study indicates the relative balance of monocytic MDSCs and antitumor immunity cells (especially CTLs, NK cells and γδT cells) may involve in maintaining tumor cells in a state of immune-mediated dormancy. In addition, high percentages of Treg cells and PMN-MDSCs are associated with the tumor immune escape state - mice with progressing sarcomas. In summary, the relative balance of immune effector cells and suppressive populations in the tumor microenvironment may involve in determining the fate of tumors.
Product Description
Supelco
3-Methylcholanthrene, analytical standard
Sigma-Aldrich
3-Methylcholanthrene, 98%