Merck
  • Enantioselective synthesis of benzofurans and benzoxazines via an olefin cross-metathesis-intramolecular oxo-Michael reaction.
Enantioselective synthesis of benzofurans and benzoxazines via an olefin cross-metathesis-intramolecular oxo-Michael reaction.
Chemical communications (Cambridge, England) (2013-07-26)
Jun-Wei Zhang, Quan Cai, Qing Gu, Xiao-Xin Shi, Shu-Li You
ABSTRACT
Chiral phosphoric acid and Hoveyda-Grubbs II were found to catalyze an olefin cross-metathesis-intramolecular oxo-Michael cascade reaction of the ortho-allylphenols and enones to provide a variety of benzofuran and benzoxazine derivatives in moderate to good yields and enantioselectivity.
Sigma-Aldrich
Phosphoric acid, JIS special grade, ≥85.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, SAJ first grade, ≥85.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.