Merck
  • Functional genomic assessment of phosgene-induced acute lung injury in mice.
Functional genomic assessment of phosgene-induced acute lung injury in mice.
American journal of respiratory cell and molecular biology (2013-04-18)
George D Leikauf, Vincent J Concel, Kiflai Bein, Pengyuan Liu, Annerose Berndt, Timothy M Martin, Koustav Ganguly, An Soo Jang, Kelly A Brant, Richard A Dopico, Swapna Upadhyay, Clinton Cario, Y P Peter Di, Louis J Vuga, Emrah Kostem, Eleazar Eskin, Ming You, Naftali Kaminski, Daniel R Prows, Daren L Knoell, James P Fabisiak
ABSTRACT
In this study, a genetically diverse panel of 43 mouse strains was exposed to phosgene and genome-wide association mapping performed using a high-density single nucleotide polymorphism (SNP) assembly. Transcriptomic analysis was also used to improve the genetic resolution in the identification of genetic determinants of phosgene-induced acute lung injury (ALI). We prioritized the identified genes based on whether the encoded protein was previously associated with lung injury or contained a nonsynonymous SNP within a functional domain. Candidates were selected that contained a promoter SNP that could alter a putative transcription factor binding site and had variable expression by transcriptomic analyses. The latter two criteria also required that ≥10% of mice carried the minor allele and that this allele could account for ≥10% of the phenotypic difference noted between the strains at the phenotypic extremes. This integrative, functional approach revealed 14 candidate genes that included Atp1a1, Alox5, Galnt11, Hrh1, Mbd4, Phactr2, Plxnd1, Ptprt, Reln, and Zfand4, which had significant SNP associations, and Itga9, Man1a2, Mapk14, and Vwf, which had suggestive SNP associations. Of the genes with significant SNP associations, Atp1a1, Alox5, Plxnd1, Ptprt, and Zfand4 could be associated with ALI in several ways. Using a competitive electrophoretic mobility shift analysis, Atp1a1 promoter (rs215053185) oligonucleotide containing the minor G allele formed a major distinct faster-migrating complex. In addition, a gene with a suggestive SNP association, Itga9, is linked to transforming growth factor β1 signaling, which previously has been associated with the susceptibility to ALI in mice.