Merck
  • Structure-based design, synthesis, and study of pyrazolo[1,5-a][1,3,5]triazine derivatives as potent inhibitors of protein kinase CK2.
Structure-based design, synthesis, and study of pyrazolo[1,5-a][1,3,5]triazine derivatives as potent inhibitors of protein kinase CK2.
Bioorganic & medicinal chemistry letters (2007-06-02)
Zhe Nie, Carin Perretta, Philip Erickson, Stephen Margosiak, Robert Almassy, Jia Lu, April Averill, Kraig M Yager, Shaosong Chu
ABSTRACT
The structure-based design, synthesis, and anticancer activity of novel inhibitors of protein kinase CK2 are described. Using pyrazolo[1,5-a][1,3,5]triazine as the core scaffold, a structure-guided series of modifications provided pM inhibitors with microM-level cytotoxic activity in cell-based assays with prostate and colon cancer cell lines.
Sigma-Aldrich
Ethoxycarbonyl isothiocyanate, 98%
Pricing and availability is not currently available.