Merck
  • A novel aryl-hydrazide from the marine lichen Lichina pygmaea: isolation, synthesis of derivatives, and cytotoxicity assays.
A novel aryl-hydrazide from the marine lichen Lichina pygmaea: isolation, synthesis of derivatives, and cytotoxicity assays.
Bioorganic & medicinal chemistry letters (2010-06-24)
Catherine Roullier, Marylène Chollet-Krugler, Pierre van de Weghe, Françoise Lohézic-Le Devehat, Joël Boustie
ABSTRACT
A new aryl-hydrazide l-glutamic acid derivative, pygmeine (3), was isolated from a methanolic extract of Lichina pygmaea, a marine lichen. Synthetic derivatives obtained via a two-step coupling of l-glutamic acid with phenylhydrazine moieties were useful to elucidate the structure of 3 and to carry out biological assays. Thus, the cytotoxicity of the ortho-, meta-, and para-hydroxyl isomers along with their respective benzyl intermediates, and a natural methoxylated analog, were evaluated on murine and human melanoma cells (B16, A375). The para-hydroxyl isomer 6 was found to be the most active (IC(50)=1.6 microM) on B16 cells.