- Speciation without chromatography using selective hydride generation: inorganic arsenic in rice and samples of marine origin.
Speciation without chromatography using selective hydride generation: inorganic arsenic in rice and samples of marine origin.
Because of the toxicity of inorganic arsenic (iAs), only iAs needs to be monitored in food and feedstuff. This demands the development of easy and quick analytical methods to screen large number of samples. This work focuses on hydride generation (HG) coupled with an ICPMS as an arsenic detector where the HG is added as a selective step to determine iAs in the gaseous phase while organically bound As remains in the solution. iAs forms volatile arsine species with high efficiency when treated with NaBH4 at acidic conditions, whereas most other organoarsenic compounds do not form any or only less volatile arsines. Additionally, using high concentrations of HCl further reduces the production of the less volatile arsines and iAs is almost exclusively formed, therefore enabling to measure iAs without a prior step of species separation using chromatography. Here, we coupled a commercially available HG system to an ICPMS and optimized for determination of iAs in rice and samples of marine origin using different acid concentrations, wet and dry plasma conditions, and different reaction gas modes. Comparing this method to conventional HPLC-ICPMS, no statistical difference in iAs concentration was found and comparable limits of detections were achieved using less than half the instrument time.