Merck
  • Efficient Stille cross-coupling reaction catalyzed by the Pd(OAc)2/Dabco catalytic system.
Efficient Stille cross-coupling reaction catalyzed by the Pd(OAc)2/Dabco catalytic system.
The Journal of organic chemistry (2005-03-25)
Jin-Heng Li, Yun Liang, De-Ping Wang, Wen-Jie Liu, Ye-Xiang Xie, Du-Lin Yin
ABSTRACT
[reaction: see text] An efficient Pd(OAc)2/Dabco-catalyzed Stille cross-coupling reaction procedure has been developed. In the presence of Pd(OAc)2 and Dabco (triethylenediamine), various aryl halides including aryl iodides, aryl bromides, and activated aryl chlorides were coupled efficiently with organotin compounds to afford the corresponding biaryls, alkene, and alkynes in good to excellent yields. Furthermore, high TONs [turnover numbers, up to 980,000 TONs for the coupling reaction of 1-bromo-4-nitrobenzene and furan-2-yltributyltin] for the Stille cross-coupling reaction were observed.
Sigma-Aldrich
1-Bromo-4-nitrobenzene, 99%
Pricing and availability is not currently available.