Merck
  • Recyclable Mitsunobu reagents: catalytic Mitsunobu reactions with an iron catalyst and atmospheric oxygen.
Recyclable Mitsunobu reagents: catalytic Mitsunobu reactions with an iron catalyst and atmospheric oxygen.
Angewandte Chemie (International ed. in English) (2013-03-08)
Daisuke Hirose, Tsuyoshi Taniguchi, Hiroyuki Ishibashi
ABSTRACT
Aerobic recycling: A catalytic amount of a hydrazine reagent is sufficient to promote Mitsunobu reactions in the presence of triphenylphosphine, an iron catalyst, and air. The active form of the catalyst, an azo species, can be readily generated by iron-catalyzed aerobic oxidation. MS = molecular sieves, Pc = phthalocyanine.
Sigma-Aldrich
3,4-Dichlorophenylhydrazine hydrochloride, technical grade, 90%
Pricing and availability is not currently available.