Merck
  • Spectrofluorimetric flow injection method for the individual and successive determination of L-cysteine and L-cystine in pharmaceutical and urine samples.
Spectrofluorimetric flow injection method for the individual and successive determination of L-cysteine and L-cystine in pharmaceutical and urine samples.
The Analyst (1992-06-01)
T Pérez-Ruiz, C Martínez-Lozano, V Tomás, J Carpena
ABSTRACT
A flow injection configuration is proposed for the determination of L-cysteine and L-cystine individually and for mixtures of both analytes. The procedure is based on the rapid oxidation of L-cysteine by thallium(III) with concomitant formation of fluorescent thallium(I). The inclusion of a selecting valve and of a copper-coated cadmium column in the configuration allows the successive determination of two analytes. Linear calibration graphs were obtained between 5 x 10(-6) and 5 x 10(-5) mol dm-3 of L-cysteine and between 2 x 10(-6) and 2 x 10(-5) mol dm-3 of L-cystine. The applicability of the method to the determination of L-cysteine and L-cystine in pharmaceutical preparations was demonstrated by investigating the effect of potential interferents and by the analysis of commercial preparations. The method was successfully applied to the determination of L-cysteine and L-cystine in urine samples.
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Pricing and availability is not currently available.