Merck
  • p-Trifluoromethyldiazirinyl-etomidate: a potent photoreactive general anesthetic derivative of etomidate that is selective for ligand-gated cationic ion channels.
p-Trifluoromethyldiazirinyl-etomidate: a potent photoreactive general anesthetic derivative of etomidate that is selective for ligand-gated cationic ion channels.
Journal of medicinal chemistry (2010-08-14)
S Shaukat Husain, Deirdre Stewart, Rooma Desai, Ayman K Hamouda, S Guo-Dong Li, Elizabeth Kelly, Zuzana Dostalova, Xiaojuan Zhou, Joseph F Cotten, Douglas E Raines, Richard W Olsen, Jonathan B Cohen, Stuart A Forman, Keith W Miller
ABSTRACT
We synthesized the R- and S-enantiomers of ethyl 1-(1-(4-(3-((trifluoromethyl)-3H-diazirin-3-yl)phenyl)ethyl)-1H-imidazole-5-carboxylate (trifluoromethyldiazirinyl-etomidate), or TFD-etomidate, a novel photoactivable derivative of the stereoselective general anesthetic etomidate (R-(2-ethyl 1-(phenylethyl)-1H-imidazole-5-carboxylate)). Anesthetic potency was similar to etomidate's, but stereoselectivity was reversed and attenuated. Relative to etomidate, TFD-etomidate was a more potent inhibitor of the excitatory receptors, nAChR (nicotinic acetylcholine receptor) ((alpha1)(2)beta1delta1gamma1) and 5-HT(3A)R (serotonin type 3A receptor), causing significant inhibition at anesthetic concentrations. S- but not R-TFD-etomidate enhanced currents elicited from inhibitory alpha1beta2gamma2L GABA(A)Rs by low concentrations of GABA, but with a lower efficacy than R-etomidate, and site-directed mutagenesis suggests they act at different sites. [(3)H]TFD-etomidate photolabeled the alpha-subunit of the nAChR in a manner allosterically regulated by agonists and noncompetitive inhibitors. TFD-etomidate's novel pharmacology is unlike that of etomidate derivatives with photoactivable groups in the ester position, which behave like etomidate, suggesting that it will further enhance our understanding of anesthetic mechanisms.
Product Description
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O