Merck
Fluorous synthesis of disubstituted pyrimidines.
Organic letters (2003-03-28)
Wei Zhang
ABSTRACT
[reaction: see text] The fluorous synthesis of disubstituted pyrimidines is carried out by attaching 2,4-dichloro-6-methylpyrimidine with 1H,1H,2H,2H-perfluorodecanethiol. The tagged substrate is substituted with 3-(trifluoromethyl)pyrazole followed by thioether oxidation and tag displacement with amines or thiols. The fluorous chain serves as a phase tag for intermediate and product purification over FluoroFlash SPE cartridges.
Sigma-Aldrich
2,4-Dichloro-6-methylpyrimidine, 98%