- KLF1 regulates BCL11A expression and gamma- to beta-globin gene switching.
KLF1 regulates BCL11A expression and gamma- to beta-globin gene switching.
We show that knockdown of KLF1 in human and mouse adult erythroid progenitors markedly reduces BCL11A levels and increases human gamma-globin/beta-globin expression ratios. These results suggest that KLF1 controls globin gene switching by directly activating beta-globin and indirectly repressing gamma-globin gene expression. Controlled knockdown of KLF1 in adult erythroid progenitors may provide a method to activate fetal hemoglobin expression in individuals with beta-thalassemia or sickle cell disease.