Merck
Molecular basis of coiled-coil formation.
Proceedings of the National Academy of Sciences of the United States of America (2007-04-18)
Michel O Steinmetz, Ilian Jelesarov, William M Matousek, Srinivas Honnappa, Wolfgang Jahnke, John H Missimer, Sabine Frank, Andrei T Alexandrescu, Richard A Kammerer
ABSTRACT
Coiled coils have attracted considerable interest as design templates in a wide range of applications. Successful coiled-coil design strategies therefore require a detailed understanding of coiled-coil folding. One common feature shared by coiled coils is the presence of a short autonomous helical folding unit, termed "trigger sequence," that is indispensable for folding. Detailed knowledge of trigger sequences at the molecular level is thus key to a general understanding of coiled-coil formation. Using a multidisciplinary approach, we identify and characterize here the molecular determinants that specify the helical conformation of the monomeric early folding intermediate of the GCN4 coiled coil. We demonstrate that a network of hydrogen-bonding and electrostatic interactions stabilize the trigger-sequence helix. This network is rearranged in the final dimeric coiled-coil structure, and its destabilization significantly slows down GCN4 leucine zipper folding. Our findings provide a general explanation for the molecular mechanism of coiled-coil formation.
Sigma-Aldrich
Anti-CCDC47 antibody produced in rabbit, affinity isolated antibody