Merck
Determination of solanidine in human plasma by radioimmunoassay.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1983-10-01)
J A Matthew, M R Morgan, R McNerney, H W Chan, D T Coxon
ABSTRACT
A radioimmunoassay for solanidine, the major hydrolysis product of potato glycoalkaloids, has been established and validated for application to human plasma. All 34 samples of human plasma tested contained solanidine, with a mean of 1.27 +/- 0.97 ng/ml (3.18 +/- 2.43 nmol/litre).
Sigma-Aldrich
Solanidine, ≥97.0% (HPLC)
Pricing and availability is not currently available.