Merck
Hyperventilation in severe diabetic ketoacidosis.
Pediatric critical care medicine : a journal of the Society of Critical Care Medicine and the World Federation of Pediatric Intensive and Critical Care Societies (2005-06-29)
Robert C Tasker, Daniel Lutman, Mark J Peters
ABSTRACT
To explore whether the carbon dioxide-bicarbonate (P(CO(2))-HCO(3)) buffering system in blood and cerebrospinal fluid (CSF) in diabetic ketoacidosis should influence the approach to ventilation in patients at risk of cerebral edema. Medline search, manual search of references in articles found in Medline search, and use of historical literature from 1933 to 1967. A clinical vignette is used--a child with severe diabetic ketoacidosis who presented with profound hypocapnia and then deteriorated--as a basis for discussion of integrative metabolic and vascular physiology. Studies included reports in diabetic ketoacidosis where arterial and CSF acid-base data have been presented. Studies where simultaneous acid-base, ventilation, respiratory quotient, and cerebral blood flow data are available. We revisit a hypothesis and, by reassessing data, put forward an argument based on the significance of low [HCO(3)](CSF) and rising Pa(CO(2))- hyperventilation in diabetic ketoacidosis and the limit in biology of survival; repair of severe diabetic ketoacidosis and Pa(CO(2))-and mechanical ventilation. The review highlights a potential problem with mechanical ventilation in severe diabetic ketoacidosis and suggests that the P(CO(2))--HCO(3) hypothesis is consistent with data on cerebral edema in diabetic ketoacidosis. It also indicates that the recommendation to avoid induced hyperventilation early in the course of intensive care may be counter to the logic of adaptive physiology.
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Supelco
1-Butanol, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%