Merck
  • Plasma and urine glycolate assays for differentiating the hyperoxaluria syndromes.
Plasma and urine glycolate assays for differentiating the hyperoxaluria syndromes.
The Journal of urology (1992-09-01)
M Marangella, M Petrarulo, C Vitale, D Cosseddu, F Linari
ABSTRACT
To differentiate hyperoxaluria syndromes we measured plasma and urine glycolate by a novel high performance liquid chromatographic procedure. Mean glycolate level was 7.9 +/- 2.4 mumol./l. in plasma and 422 +/- 137 mumol./24 hours in urine from 19 control subjects. Renal clearance was about 50% the glomerular filtration rate irrespective of the underlying disease. There was close correlation between glycolate and oxalate in plasma. Plasma glycolate was normal in all but 8 patients who had primary hyperoxaluria 1. Plasma assay detected the disease more efficiently than urine assay. Pyridoxine decreased oxalate biosynthesis in 2 of the 4 patients treated with it and glycolate assay confirmed this behavior. Glycolate excretion was significantly high in 3 of 8 patients of primary hyperoxaluria 1 patients. Idiopathic stone formers had mild increases in glycolate excretion but this was not related with oxalate excretion. Glycolate levels were normal in 5 patients with enteric hyperoxaluria. We conclude that glycolate assay is essential for identifying patients with primary hyperoxaluria 1 and may represent a valuable tool for differentiating hyperoxaluria.
Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%
Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%