Merck
  • Cyanidin 3-glucoside and peonidin 3-glucoside inhibit tumor cell growth and induce apoptosis in vitro and suppress tumor growth in vivo.
Cyanidin 3-glucoside and peonidin 3-glucoside inhibit tumor cell growth and induce apoptosis in vitro and suppress tumor growth in vivo.
Nutrition and cancer (2006-04-01)
Pei-Ni Chen, Shu-Chen Chu, Hui-Ling Chiou, Chui-Liang Chiang, Shun-Fa Yang, Yih-Shou Hsieh
ABSTRACT
Dietary polyphenols, including anthocyanins, are suggested to be involved in the protective effects of fruits and vegetables against cancer. However, anticancer effects of peonidin 3-glucoside have not been clearly demonstrated, with only limited studies being available concerning the inhibitory effect of cyanidin 3-glucoside for tumor cell growth. Therefore, in this study, we have isolated and identified the two bioactive compounds, peonidin 3-glucoside and cyanidin 3-glucoside, from Oryza sativa L. indica, to treat various cancer cells. The results showed that, among analyzed cell lines, HS578T was the most sensitive to peonidin 3-glucoside and cyanidin 3-glucoside. Treatment with peonidin 3-glucoside or cyanidin 3-glucoside resulted in a strong inhibitory effect on cell growth via G2/M arrest. Regarding cell cyclerelated proteins, peonidin 3-glucoside treatment resulted in down-regulation of protein levels of cyclin-dependent kinase (CDK)-1, CDK-2, cyclin B1, and cyclin E, whereas cyanidin 3-glucoside could decrease the protein levels of CDK-1, CDK-2, cyclin B1, and cyclin D1. In addition, cyanidin 3-glucoside or peonidin 3-glucoside also induced caspase-3 activation, chromatin condensation, and cell death. Furthermore, anthocyanins from O. sativa L. indica were evidenced by their inhibition on the growth of Lewis lung carcinoma cells in vivo.
Sigma-Aldrich
Peonidin 3-O-glucoside chloride, ≥95% (HPLC)
Pricing and availability is not currently available.