Merck
  • On the analysis of 1-nitronaphthalene, 1-nitropyrene and 6-nitrochrysene in cigarette smoke.
On the analysis of 1-nitronaphthalene, 1-nitropyrene and 6-nitrochrysene in cigarette smoke.
Carcinogenesis (1985-04-01)
K el-Bayoumy, M O'Donnell, S S Hecht, D Hoffmann
ABSTRACT
A method was developed for the analysis of 1-nitronaphthalene, 1-nitropyrene, and 6-nitrochrysene in mainstream cigarette smoke, using [14C]1-nitronaphthalene, [14C]1-nitropyrene, and [14C]6-nitrochrysene as tracers and internal standards. Cigarette smoke condensate was collected and the appropriate fractions containing the labelled standards were obtained by a series of solvent partitions and column chromatography. Recovery of internal standards ranged from 60 to 70%. The fractions were analyzed by capillary gas chromatography with electron capture detection and by combined capillary gas chromatography-mass spectrometry. 1-Nitronaphthalene (less than 10 ng/cigarette), 1-nitropyrene (less than 10 ng/cigarette), and 6-nitrochrysene (less than 1 ng/cigarette) were not detected in the mainstream smoke of the University of Kentucky 1R1 cigarette, a US commercial 85 mm non-filter cigarette, or a French commercial 70 mm non-filter cigarette.
Sigma-Aldrich
1-Nitronaphthalene, 99%
Pricing and availability is not currently available.