Merck
Detection of DNA-crosslinking agents with the alkaline comet assay.
Environmental and molecular mutagenesis (1996-01-01)
S Pfuhler, H U Wolf
ABSTRACT
The single cell gel electrophoresis, or comet assay, under alkaline conditions is a sensitive, simple and rapid method for the detection of DNA damage at the individual cell level. Its applicability as an indicator for the DNA crosslinking potency of a test substance was investigated in human white blood cells by combined treatment with the DNA damaging agent methyl methanesulphonate (MMS) for 2 hr at 37 degrees C. The known crosslinking agents cisplatinum, mitomycin C and formaldehyde, and the formaldehyde releasers diazolidinyl urea and dimethylol urea, were shown to reduce MMS-induced DNA migration in the comet assay in a concentration-dependent manner. Two other protocols, adding MMS to the cells before or after treatment with a crosslinking agent, were carried out and achieved similar results. The results of this study indicate that the comet assay is a useful tool for the detection of crosslinking agents. Advantages and limitations of this method compared to the alkaline elution technique are discussed.
Sigma-Aldrich
N,N′-Bis(hydroxymethyl)urea, ≥95%
Pricing and availability is not currently available.