Merck
  • Simultaneous determination of dihydrocodeine and dihydromorphine in serum by gas chromatography-tandem mass spectrometry.
Simultaneous determination of dihydrocodeine and dihydromorphine in serum by gas chromatography-tandem mass spectrometry.
Journal of chromatography. B, Biomedical applications (1995-01-06)
U Hofmann, M F Fromm, S Johnson, G Mikus
ABSTRACT
A sensitive and specific method was developed for the determination of dihydrocodeine and its metabolite dihydromorphine in human serum using codeine and morphine as internal standards. Measurement is performed with GC-tandem MS after one simple extraction step and derivatization to the pentafluoropropionic esters. Sensitivity of the method is excellent and allows for the reproducible quantification of dihydrocodeine and dihydromorphine with limits of quantification of 2 ng/ml and 40 pg/ml serum, respectively. The method is therefore well suited for investigation of the pharmacokinetics and the metabolism of dihydrocodeine.