Merck
[Acute toxicity of two phenylcarbamates: propham and chloropropham].
Comptes rendus des seances de la Societe de biologie et de ses filiales (1981-01-01)
B Caporiccio, J Tournamille, R Michel
ABSTRACT
The bibliographic data dealing with the acute toxicity of two phenylcarbamates, propham and chloropropham, are not very precise an often contradictory. This work is a restatement and also a demonstration of the high sensitivity of the aquatic animals that we have tested (Amphibia and Crustacea) in comparison with the Mammals (rat and mouse).
Product Description
Supelco
Propham, PESTANAL®, analytical standard
Pricing and availability is not currently available.