Merck
  • Inhibitors of poly(ADP-ribose) synthesis enhance X-ray killing of log-phase Chinese hamster cells.
Inhibitors of poly(ADP-ribose) synthesis enhance X-ray killing of log-phase Chinese hamster cells.
Radiation research (1984-03-01)
E Ben-Hur, H Utsumi, M M Elkind
ABSTRACT
Postirradiation incubation of V79 Chinese hamster cells with inhibitors of poly(ADP-ribose) synthesis was found to potentiate the killing of cells by X rays. Potentiation increased with incubation time and with concentration of the inhibitor. Preirradiation incubation had only a small effect. The enhanced response correlated well with the known extent of the inhibition of poly(ADP-ribose) synthesis. A radiation-sensitive line, V79- AL162 /S-10, was affected to a lesser extent than the normal cells. Cells repaired the radiation damage with which the inhibitors interacted within 1 hr, a process that has similar kinetics to what is observed when a postirradiation treatment with hypertonic buffer is used [H. Utsumi and M. M. Elkind , Radiat . Res. 77, 346-360 (1979)]. However, the sectors of damage affected by inhibitors of poly(ADP-ribose) synthesis and hypertonic buffer do not entirely overlap. The inhibitor nicotinamide enhanced the killing mainly of late S-phase cells and did not affect cells at the G1/S border. It is concluded that the repair process(es) involving poly(ADP-ribose) synthesis is important for cell survival in repair-competent cells and that the radiation-sensitive cells that were examined are partially deficient in a repair pathway in which poly(ADP-ribose) participates.
Sigma-Aldrich
4-Aminobenzamide, 98%
Pricing and availability is not currently available.