Merck
  • Seasonal peculiarities of thyroxine influence on the way of serotonin metabolism in the pineal gland of the rat.
Seasonal peculiarities of thyroxine influence on the way of serotonin metabolism in the pineal gland of the rat.
Acta physiologica Hungarica (1987-01-01)
E S Rom-Bugoslavskaja, L Bondarenko
ABSTRACT
In intact Wistar pubertal male rats held on LD 8:16 (winter) and 16:8 (summer) pineal melatonin (M) production and other pineal indoles content serotonin (S), 5-methoxytriptamine (5-MT), N-acetylserotonin (N-aS), 5-hydroxyindoleacetic acid (5-HIAA) and 5-methoxyindoleacetic acid (5-MIAA) were investigated in basic conditions and after 10-days T4 administration. The results suggest, that intact control in winter comparing with summer demonstrates high M, N-aS, 5-HIAA and 5-MIAA level, but low 5-MT concentrations, S level was not changed. T4 administration in winter produced a pronounced augmentation of all parameters (excluding serotonin), in summer M production not only increased, but appears to be dependent on the active metabolism of his precursors (S, 5-MT, N-aS). The experimental data favour more profound comprehension to the mechanisms of pineal activity regulation.
Sigma-Aldrich
5-Methoxy-3-indoleacetic acid, 98%