Merck
  • Royal sun medicinal mushroom Agaricus brasiliensis (higher Basidiomycetes) and the attenuation of pulmonary inflammation induced by 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK).
Royal sun medicinal mushroom Agaricus brasiliensis (higher Basidiomycetes) and the attenuation of pulmonary inflammation induced by 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK).
International journal of medicinal mushrooms (2013-06-26)
Carolina Croccia, Agnaldo Jose Lopes, Luis Felipe Ribeiro Pinto, Armando Ubirajara Oliveira Sabaa-Srur, Luiz Carlos Aguiar Vaz, Marcele Nogueira de Sousa Trotte, Bernardo Tessarollo, Aristofanes Correa Silva, Haroldo Jose de Matos, Rodolfo Acatauassu Nunes
ABSTRACT
Agaricus brasiliensis currently is one of the most studied fungi because of its nutritional and therapeutic properties as an anti-inflammatory agent and an adjuvant in cancer chemotherapy. The effects of orally administered aqueous A. brasiliensis extract (14.3- and 42.9-mg doses) on parenchymal lung damage induced by carcinogenic 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) were observed in Wistar rats. NNK treatment induced pulmonary inflammation, but not lung cancer, in the rats. The lungs of animals treated with NNK showed a higher level of inflammation than those of the control group according to histopathologic examinations (P < 0.01) and kurtosis analysis (P < 0.001) of a global histogram generated from thoracic computed tomography scans. There was no significant difference in the alveolar and bronchial exudates between animals treated with a 14.3-mg dose of A. brasiliensis extract and the control without NNK. However, a significant difference was found between animals treated with NNK, received a 42.9-mg dose of A. brasiliensis (P < 0.05), and the controls not treated with NNK. We did not observe a significant difference between the kurtoses of the A. brasiliensis (14.3 mg) and control groups. However, a 42.9-mg dose of A. brasiliensis resulted in lower kurtosis values than those observed in the control group (P < 0.001). In conclusion, a low dose of A. brasiliensis was more effective in attenuating pulmonary inflammation. Similar to the histopathological results, the computed tomography scans also showed a protective effect of A. brasiliensis at the lower dose, which prevented gross pulmonary consolidation.
Product Description
Supelco
4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone, analytical standard
Pricing and availability is not currently available.
Supelco
4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.