Merck
  • [Evaluation of the treatment effectiveness of domestic G-SCF preparations in experiments on irradiated dogs].
[Evaluation of the treatment effectiveness of domestic G-SCF preparations in experiments on irradiated dogs].
Radiatsionnaia biologiia, radioecologiia (2013-05-25)
L M Rozhdestvenskiĭ, T G Shliakova, R A Shchegoleva, N I Lisina, V V Zorin
ABSTRACT
We have evaluated the treatment effectiveness of Leucostim and Neupomax in dogs exposed to radiation at lethal doses of 3 and 3.5 Gy, correspondingly, by testing the dynamics of the blood cell number, first of all, leucocytes and neutrophiles, and the 45-day survival. Supportive therapy for all the dogs, including the control ones, consisted in antibiotic treatment during the acute period of 7-24 days. It was shown that both pre-parations administered consecutively for about 17-21 days after irradiation positively influenced the dynamics of all blood cells but predominantly impacted the neutrophile number dynamics. The latter ones manifested a higher nadir level and an earlier onset of restoration in the G-SCF treated dogs in comparison with the control ones. The tendency to a positive influence on the survival has been shown in Neupomax-treated dogs exposed to 3.5 Gy of radiation (plus about 40%). The results of the experiments were in good accordance with the data by foreign authors who used Neupogen. This allows a conclusion that home-produced G-SCF preparations can replace their foreign analogues.