Merck
  • Copper-catalyzed synthesis of substituted benzothiazoles via condensation of 2-aminobenzenethiols with nitriles.
Copper-catalyzed synthesis of substituted benzothiazoles via condensation of 2-aminobenzenethiols with nitriles.
Organic letters (2013-03-19)
Yadong Sun, Huanfeng Jiang, Wanqing Wu, Wei Zeng, Xia Wu
ABSTRACT
An efficient and convenient method was developed for the formation of 2-substituted benzothiazoles via a copper-catalyzed condensation of 2-aminobenzenethiols with nitriles. The developed method is applicable to a wide range of nitriles containing different functional groups furnishing excellent yields of the corresponding products.
Sigma-Aldrich
Sulfanilic acid, JIS special grade, 99.0-100.5%
Sigma-Aldrich
Sulfanilic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sulfanilic acid, ACS reagent, 99%
Millipore
Nitrate Reagent B, suitable for microbiology