- Dissecting glycosylation steps in lobophorin biosynthesis implies an iterative glycosyltransferase.
Dissecting glycosylation steps in lobophorin biosynthesis implies an iterative glycosyltransferase.
Organic letters (2013-02-26)
Sumei Li, Ji Xiao, Yiguang Zhu, Guangtao Zhang, Chunfang Yang, Haibo Zhang, Liang Ma, Changsheng Zhang
PMID23432710
ABSTRACT
The identification of a lobophorin biosynthetic gene cluster from the deep-sea derived Streptomyces sp. SCSIO 01127 reveals a paradigm of three glycosyltransferases (GTs) LobG1-LobG3 being responsible for appending four sugars. Characterization of five differentially glycosylated metabolites from three GT gene-inactivation mutants allowed the assignment of GT functions and the implication of LobG3 as an iterative GT to attach two digitoxoses.