Merck
  • A randomized, non-inferiority study comparing efficacy and safety of a single dose of pegfilgrastim versus daily filgrastim in pediatric patients after autologous peripheral blood stem cell transplant.
A randomized, non-inferiority study comparing efficacy and safety of a single dose of pegfilgrastim versus daily filgrastim in pediatric patients after autologous peripheral blood stem cell transplant.
PloS one (2013-01-12)
Simone Cesaro, Francesca Nesi, Gloria Tridello, Massimo Abate, Irene Sara Panizzolo, Rita Balter, Elisabetta Calore
ABSTRACT
To assess the non-inferiority of pegfilgrastim versus filgrastim in speeding the recovery of polymorphonuclear cells (PMN) in pediatric patients who underwent autologous peripheral blood stem cell transplant (PBSCT). The sample size of this randomized, multicenter, phase III study, was calculated assuming that a single dose of pegfilgrastim of 100 ug/kg was not inferior to 9 doses of filgrastim of 5 ug/kg/day. Randomization was performed by a computer-generated list and stored by sequentially numbered sealed envelopes. Sixty-one patients, with a median age of 11.5 years, were recruited: 29 in the filgrastim arm and 32 in the pegfilgrastim arm. Twenty percent were affected by lymphoma/leukaemia and eighty percent by solid tumors. The mean time to PMN engraftment was 10.48 days (standard deviation [SD] 1.57) and 10.44 days (SD 2.44) in the filgrastim and pegfilgrastim arms, respectively. Having fixed a non-inferiority margin Delta of 3, the primary endpoint of non-inferiority was reached. No differences were observed for other secondary endpoints: platelet engraftment, mean time to platelet recovery (28 days vs. 33 days), fever of unknown origin (79% vs. 78%), proven infection (34% vs. 28%), mucositis (76% vs. 59%). After a median follow-up of 2.3 years (95% C.I.: 1.5, 3.3), 20 deaths were observed due to disease progression. We conclude that pegfilgrastim was not inferior to daily filgrastim in pediatric patients who underwent PBSCT. EU CLINICAL TRIAL REGISTER NUMBER: 2007-001430-14.