Merck
  • Release of avian neurotensin in response to intraluminal contents in the duodenum of chickens.
Release of avian neurotensin in response to intraluminal contents in the duodenum of chickens.
Poultry science (2013-01-10)
Teresa F DeGolier, Robert E Carraway, Gary E Duke
ABSTRACT
Peripheral and hepatic-portal plasma levels of neurotensin (NT) in fed and fasted chickens were determined using RIA. Portal levels of NT(1-13) (fed = 61.3 ± 3.9 fmol/mL; fasted = 44.5 ± 3.9 fmol/mL) were significantly higher than peripheral levels (fed = 8.2 ± 3.3 fmol/mL; fasted = 7.8 ± 3.0 fmol/mL) collected from the wing vein, indicating that some NT is metabolized in the liver. Portal plasma levels of NT collected from fed birds were also significantly higher than portal plasma levels of NT collected from fasted birds. Neurotensin, as identified by HPLC, exhibited a 2-fold increase in plasma extracts following perfusion of the proximal ileum with a 10-mg sample of oleic acid, as compared with control samples of plasma collected before oleic acid perfusion. In whole-animal studies, the injection of a micellar solution of oleic acid into isolated segments of the duodenum resulted in elevated plasma immunoreactive NT in blood collected from the pancreaticoduodenal vein. Injection of a 1,000 mOsm sodium chloride solution had a slightly lesser and delayed effect compared with oleic acid, but a greater effect than 0.1 N hydrochloric acid in isotonic saline solution. Injection of an amino acid solution (10% Travasol), 300 mOsm glucose solution, or pure corn oil had no effect. These results demonstrate that intraduodenal oleic acid is a potent stimulus for the release of NT from the duodenum into the hepatic-portal circulation of chickens.
Product Description
Supelco
Corn oil, analytical standard
Sigma-Aldrich
Corn oil, CP
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds