Merck
  • Enantioselective decarboxylative amination: synthesis of axially chiral allenyl amines.
Enantioselective decarboxylative amination: synthesis of axially chiral allenyl amines.
Angewandte Chemie (International ed. in English) (2012-09-11)
Baoqiang Wan, Shengming Ma
ABSTRACT
Getting axed: synthesis of the title amines, bearing functionality (R(1) and R(2)), involves the enantioselective palladium-catalyzed decarboxylation of allenyl N-tosylcarbamates. The reaction proceeds smoothly using both the chiral ligands (S)- and (R)-DTBM-Segphos (1) to afford the allenyl amines in good yields and with high enantioseletivities.