Merck
  • Analysis of the properties of dental cements after exposure to incubation media containing Streptococcus mutans.
Analysis of the properties of dental cements after exposure to incubation media containing Streptococcus mutans.
The journal of contemporary dental practice (2012-01-25)
Fernando Carlos Hueb de Menezes, Geraldo Thedei Junior, Wildomar Jose de Oliveira, Tony de Paiva Paulino, Marcelo Boaventura de Moura, Igor Lima da Silva, Marcos Boaventura de Moura
ABSTRACT
Indirect restorations are increasingly used in dentistry, and the cementation interface is possibly the most critical region of the work. The objective of the present work was to evaluate the influence of exposure to a culture medium containing S. mutans on the hardness and solubility of four different cementing agents (zinc phosphate, glass ionomer, glass ionomer modified with resin and resin cement). Test specimens composed of these cements were exposed for 30 days in a culture medium containing S. mutans. After leaching, the test materials were assessed in terms of their solubility (loss of mass) and Knoop (KHN) microhardness. Changes in surface morphology were identified using scanning electron microscopy (SEM). The resin cement showed no significant solubility and its hardness increased following exposure and leaching, while the zinc phosphate cement was the most soluble and its hardness decreased after exposure to the culture medium. SEM analyses identified morphological alterations on the surfaces of the test materials that were compatible with the solubility results. It is concluded that resinous cements perform better than water-based cements when exposed to acidic conditions. The effects of acids from Streptococcus mutans can interfere with the efficiency and properties of some cements used for fixation of indirect restorations, exposed to the buccal environment.
Sigma-Aldrich
Zinc phosphate, 99.998% trace metals basis
Pricing and availability is not currently available.