Merck
  • Stereoselective synthesis of sugar fused β-disubstituted γ-butyro-lactones: C-spiro-glycosides from 1,2-cyclopropanecarboxylated sugars.
Stereoselective synthesis of sugar fused β-disubstituted γ-butyro-lactones: C-spiro-glycosides from 1,2-cyclopropanecarboxylated sugars.
Chemical communications (Cambridge, England) (2011-12-03)
Perali Ramu Sridhar, Kalapati Seshadri, Gadi Madhusudhan Reddy
ABSTRACT
A stereoselective methodology was developed for the construction of C-spiro-glycosides in two steps involving bromonium ion activated solvolytic ring opening of sugar derived 1,2-cyclopropanecarboxylates followed by a one-pot dehydrohalogenation, intramolecular hetero-Michael addition (IHMA) and ester hydrolysis. The obtained spirocyclic lactols were further converted to enantiomerically pure spirolactones.
Sigma-Aldrich
Cyclopropanecarboxylic acid, 95%
Pricing and availability is not currently available.