Merck
  • Intensified chemotherapy with ACVBP plus rituximab versus standard CHOP plus rituximab for the treatment of diffuse large B-cell lymphoma (LNH03-2B): an open-label randomised phase 3 trial.
Intensified chemotherapy with ACVBP plus rituximab versus standard CHOP plus rituximab for the treatment of diffuse large B-cell lymphoma (LNH03-2B): an open-label randomised phase 3 trial.
Lancet (London, England) (2011-11-29)
Christian Récher, Bertrand Coiffier, Corinne Haioun, Thierry Jo Molina, Christophe Fermé, Olivier Casasnovas, Catherine Thiéblemont, André Bosly, Guy Laurent, Franck Morschhauser, Hervé Ghesquières, Fabrice Jardin, Serge Bologna, Christophe Fruchart, Bernadette Corront, Jean Gabarre, Christophe Bonnet, Maud Janvier, Danielle Canioni, Jean-Philippe Jais, Gilles Salles, Hervé Tilly
ABSTRACT
The outcome of diffuse large B-cell lymphoma has been substantially improved by the addition of the anti-CD20 monoclonal antibody rituximab to chemotherapy regimens. We aimed to assess, in patients aged 18-59 years, the potential survival benefit provided by a dose-intensive immunochemotherapy regimen plus rituximab compared with standard treatment plus rituximab. We did an open-label randomised trial comparing dose-intensive rituximab, doxorubicin, cyclophosphamide, vindesine, bleomycin, and prednisone (R-ACVBP) with subsequent consolidation versus standard rituximab, doxorubicin, cyclophosphamide, vincristine, and prednisone (R-CHOP). Random assignment was done with a computer-assisted randomisation-allocation sequence with a block size of four. Patients were aged 18-59 years with untreated diffuse large B-cell lymphoma and an age-adjusted international prognostic index equal to 1. Our primary endpoint was event-free survival. Our analyses of efficacy and safety were of the intention-to-treat population. This study is registered with ClinicalTrials.gov, number NCT00140595. One patient withdrew consent before treatment and 54 did not complete treatment. After a median follow-up of 44 months, our 3-year estimate of event-free survival was 81% (95% CI 75-86) in the R-ACVBP group and 67% (59-73) in the R-CHOP group (hazard ratio [HR] 0·56, 95% CI 0·38-0·83; p=0·0035). 3-year estimates of progression-free survival (87% [95% CI, 81-91] vs 73% [66-79]; HR 0·48 [0·30-0·76]; p=0·0015) and overall survival (92% [87-95] vs 84% [77-89]; HR 0·44 [0·28-0·81]; p=0·0071) were also increased in the R-ACVBP group. 82 (42%) of 196 patients in the R-ACVBP group experienced a serious adverse event compared with 28 (15%) of 183 in the R-CHOP group. Grade 3-4 haematological toxic effects were more common in the R-ACVBP group, with a higher proportion of patients experiencing a febrile neutropenic episode (38% [75 of 196] vs 9% [16 of 183]). Compared with standard R-CHOP, intensified immunochemotherapy with R-ACVBP significantly improves survival of patients aged 18-59 years with diffuse large B-cell lymphoma with low-intermediate risk according to the International Prognostic Index. Haematological toxic effects of the intensive regimen were raised but manageable. Groupe d'Etudes des Lymphomes de l'Adulte and Amgen.
Sigma-Aldrich
Vindesine sulfate salt hydrate, ≥95% (HPLC)